missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzhydryl Chloride 96.0+%, TCI America™

Catalog No. C1234500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1234500G 500 g
C123425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1234500G Supplier TCI America Supplier No. C1234500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 134-83-8
Molecular Formula C13H10Cl2
Molecular Weight (g/mol) 237.123
MDL Number MFCD00000856
InChI Key ALKWTKGPKKAZMN-UHFFFAOYSA-N
Synonym 4-chlorobenzhydryl chloride, 4-chlorobenzhydrylchloride, p-chlorobenzhydryl chloride, 1-chloro-4-chloro phenyl methyl benzene, benzene, 1-chloro-4-chlorophenylmethyl, chloro 4-chlorophenyl phenylmethane, chloro p-chlorophenyl phenylmethane, methane, chloro p-chlorophenyl phenyl, 1-chloro-4-chlorophenylmethyl benzene
PubChem CID 241584
IUPAC Name 1-chloro-4-[chloro(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)Cl

Specifications

Color Yellow
Boiling Point 160°C
Quantity 500 g
UN Number 1760
Formula Weight 237.12
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorobenzhydryl Chloride
EINECSNumber (9)-0387
TSCA Yes