missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzhydryl chloride, 97%

CAS: 134-83-8 | C13H10Cl2 | 237.123 g/mol

$70.97 - $584.32

Chemical Identifiers

CAS 134-83-8
Molecular Formula C13H10Cl2
Molecular Weight (g/mol) 237.123
MDL Number MFCD00000856
InChI Key ALKWTKGPKKAZMN-UHFFFAOYSA-N
Synonym 4-chlorobenzhydryl chloride, 4-chlorobenzhydrylchloride, p-chlorobenzhydryl chloride, 1-chloro-4-chloro phenyl methyl benzene, benzene, 1-chloro-4-chlorophenylmethyl, chloro 4-chlorophenyl phenylmethane, chloro p-chlorophenyl phenylmethane, methane, chloro p-chlorophenyl phenyl, 1-chloro-4-chlorophenylmethyl benzene
PubChem CID 241584
IUPAC Name 1-chloro-4-[chloro(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2277814
View Documents
Thermo Scientific Chemicals
B2277814
25 g
Each for $70.97
Only null left
 
AAB2277822
View Documents
Thermo Scientific Chemicals
B2277822
100 g
Each for $165.41
Only null left
 
AAB2277836
View Documents
Thermo Scientific Chemicals
B2277836
500 g
Each for $584.32
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

134-83-8
237.123
ALKWTKGPKKAZMN-UHFFFAOYSA-N
241584
C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)Cl
C13H10Cl2
MFCD00000856
4-chlorobenzhydryl chloride, 4-chlorobenzhydrylchloride, p-chlorobenzhydryl chloride, 1-chloro-4-chloro phenyl methyl benzene, benzene, 1-chloro-4-chlorophenylmethyl, chloro 4-chlorophenyl phenylmethane, chloro p-chlorophenyl phenylmethane, methane, chloro p-chlorophenyl phenyl, 1-chloro-4-chlorophenylmethyl benzene
1-chloro-4-[chloro(phenyl)methyl]benzene

Specifications

134-83-8
159°C to 160°C (2 mmHg)
C13H10Cl2
MFCD00000856
UN3265
Moisture sensitive
ALKWTKGPKKAZMN-UHFFFAOYSA-N
1-chloro-4-[chloro(phenyl)methyl]benzene
241584
98%
1.239
194°C (381°F)
1.603
25 g
2050236
4-chlorobenzhydryl chloride, 4-chlorobenzhydrylchloride, p-chlorobenzhydryl chloride, 1-chloro-4-chloro phenyl methyl benzene, benzene, 1-chloro-4-chlorophenylmethyl, chloro 4-chlorophenyl phenylmethane, chloro p-chlorophenyl phenylmethane, methane, chloro p-chlorophenyl phenyl, 1-chloro-4-chlorophenylmethyl benzene
C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)Cl
237.123
237.13
4-Chlorobenzhydryl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 205-158-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only