missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzhydryl chloride, 97%

Catalog No. AAB2277836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2277836 500 g
AAB2277814 25 g
AAB2277822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2277836 Supplier Thermo Scientific Chemicals Supplier No. B2277836

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 134-83-8 | C13H10Cl2 | 237.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 134-83-8
Molecular Formula C13H10Cl2
Molecular Weight (g/mol) 237.123
MDL Number MFCD00000856
InChI Key ALKWTKGPKKAZMN-UHFFFAOYSA-N
Synonym 4-chlorobenzhydryl chloride, 4-chlorobenzhydrylchloride, p-chlorobenzhydryl chloride, 1-chloro-4-chloro phenyl methyl benzene, benzene, 1-chloro-4-chlorophenylmethyl, chloro 4-chlorophenyl phenylmethane, chloro p-chlorophenyl phenylmethane, methane, chloro p-chlorophenyl phenyl, 1-chloro-4-chlorophenylmethyl benzene
PubChem CID 241584
IUPAC Name 1-chloro-4-[chloro(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)Cl

Specifications

Density 1.239
Boiling Point 159°C to 160°C (2 mmHg)
Flash Point 194°C (381°F)
Refractive Index 1.603
Quantity 500 g
UN Number UN3265
Beilstein 2050236
Sensitivity Moisture sensitive
Formula Weight 237.13
Percent Purity 98%
Chemical Name or Material 4-Chlorobenzhydryl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 205-158-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only