missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzhydrazide, 98%

CAS: 536-40-3 | C7H7ClN2O | 170.60 g/mol

$54.56 - $589.62

Chemical Identifiers

CAS 536-40-3
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.60
MDL Number MFCD00007603
InChI Key PKBGHORNUFQAAW-UHFFFAOYSA-N
Synonym 4-chlorobenzhydrazide, 4-chlorobenzoylhydrazine, 4-chlorobenzoic hydrazide, p-chlorobenzohydrazide, 4-chlorobenzoic acid hydrazide, p-chlorobenzoylhydrazine, p-chlorobenzoic hydrazide, p-chlorobenzoyl hydrazide, 4-chlorobenzoyl hydrazide, p-chlorobenzoic acid, hydrazide
PubChem CID 10816
IUPAC Name 4-chlorobenzohydrazide
SMILES NNC(=O)C1=CC=C(Cl)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1191106
View Documents
Thermo Scientific Chemicals
A1191106
5 g
Each for $54.56
Only null left
 
AAA1191118
View Documents
Thermo Scientific Chemicals
A1191118
50 g
Each for $324.82
Only null left
 
AAA1191122
View Documents
Thermo Scientific Chemicals
A1191122
100 g
Each for $589.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

536-40-3
170.60
PKBGHORNUFQAAW-UHFFFAOYSA-N
10816
NNC(=O)C1=CC=C(Cl)C=C1
C7H7ClN2O
MFCD00007603
4-chlorobenzhydrazide, 4-chlorobenzoylhydrazine, 4-chlorobenzoic hydrazide, p-chlorobenzohydrazide, 4-chlorobenzoic acid hydrazide, p-chlorobenzoylhydrazine, p-chlorobenzoic hydrazide, p-chlorobenzoyl hydrazide, 4-chlorobenzoyl hydrazide, p-chlorobenzoic acid, hydrazide
4-chlorobenzohydrazide

Specifications

536-40-3
C7H7ClN2O
5 g
4-chlorobenzhydrazide, 4-chlorobenzoylhydrazine, 4-chlorobenzoic hydrazide, p-chlorobenzohydrazide, 4-chlorobenzoic acid hydrazide, p-chlorobenzoylhydrazine, p-chlorobenzoic hydrazide, p-chlorobenzoyl hydrazide, 4-chlorobenzoyl hydrazide, p-chlorobenzoic acid, hydrazide
NNC(=O)C1=CC=C(Cl)C=C1
170.60
170.6
4-Chlorobenzhydrazide
162°C to 166°C
MFCD00007603
511154
PKBGHORNUFQAAW-UHFFFAOYSA-N
4-chlorobenzohydrazide
10816
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 208-632-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only