missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzenethiol 98.0+%, TCI America™
Supplier: TCI America C0129500G
Specifications
| 4-Chlorobenzenethiol | |
| 53°C | |
| 206°C | |
| MFCD00004847 | |
| 1759 | |
| VZXOZSQDJJNBRC-UHFFFAOYSA-N | |
| 4-chlorobenzenethiol | |
| 7815 | |
| ≥98.0% (GC,T) |
| 106-54-7 | |
| White-Yellow | |
| C6H5ClS | |
| 500 g | |
| 4-chlorothiophenol, p-chlorothiophenol, benzenethiol, 4-chloro, p-chlorobenzenethiol, p-chlorthiofenol, p-chlorophenylmercaptan, benzenethiol, p-chloro, p-mercaptochlorobenzene, 4-chlorophenylmercaptan, 1-chloro-4-mercaptobenzene | |
| C1=CC(=CC=C1S)Cl | |
| 144.616 | |
| 144.62 | |
| Crystalline Powder |
Chemical Identifiers
| 106-54-7 | |
| 144.616 | |
| VZXOZSQDJJNBRC-UHFFFAOYSA-N | |
| 7815 | |
| C1=CC(=CC=C1S)Cl |
| C6H5ClS | |
| MFCD00004847 | |
| 4-chlorothiophenol, p-chlorothiophenol, benzenethiol, 4-chloro, p-chlorobenzenethiol, p-chlorthiofenol, p-chlorophenylmercaptan, benzenethiol, p-chloro, p-mercaptochlorobenzene, 4-chlorophenylmercaptan, 1-chloro-4-mercaptobenzene | |
| 4-chlorobenzenethiol |
Safety and Handling
EINECSNumber : (3)-2901
RTECSNumber : DC1050000
TSCA : Yes