missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzenethiol 98.0+%, TCI America™

Catalog No. C012925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C012925G 25 g
C0129500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C012925G Supplier TCI America Supplier No. C012925G

Chemical Identifiers

CAS 106-54-7
Molecular Formula C6H5ClS
Molecular Weight (g/mol) 144.616
MDL Number MFCD00004847
InChI Key VZXOZSQDJJNBRC-UHFFFAOYSA-N
Synonym 4-chlorothiophenol, p-chlorothiophenol, benzenethiol, 4-chloro, p-chlorobenzenethiol, p-chlorthiofenol, p-chlorophenylmercaptan, benzenethiol, p-chloro, p-mercaptochlorobenzene, 4-chlorophenylmercaptan, 1-chloro-4-mercaptobenzene
PubChem CID 7815
IUPAC Name 4-chlorobenzenethiol
SMILES C1=CC(=CC=C1S)Cl

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 206°C
Quantity 25 g
UN Number 1759
Formula Weight 144.62
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzenethiol
EINECSNumber (3)-2901
RTECSNumber DC1050000
TSCA Yes