missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzenesulfinic acid sodium salt hydrate, 97%

Catalog No. AAA1258306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1258306 5 g
AAA1258314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1258306 Supplier Thermo Scientific Chemicals Supplier No. A1258306

CAS: 14752-66-0 | C6H4ClNaO2S | 198.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14752-66-0
Molecular Formula C6H4ClNaO2S
Molecular Weight (g/mol) 198.60
MDL Number MFCD00035602
InChI Key JFXAUUFCZJYLJF-UHFFFAOYSA-M
Synonym sodium 4-chlorobenzenesulfinate, 4-chlorobenzenesulfinic acid sodium salt, sodium 4-chlorobenzene sulfinate, unii-x2u129fz4n, sodium p-chlorobenzenesulphinate, sodium 4-chlorobenzene-1-sulfinate, 4-chlorobenzene sulfinic acid sodium, 4-chlorobenzenesulfinic acid sodium salt hydrate, p-chlorobenzenesulfinic acid, sodium salt, acmc-1cin1
PubChem CID 23664783
IUPAC Name sodium;4-chlorobenzenesulfinate
SMILES [Na+].[O-]S(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 3575604
Formula Weight 198.60 (Anhydrous)
Percent Purity 97%
Chemical Name or Material 4-Chlorobenzenesulfinic acid sodium salt hydrate
EINECSNumber 238-811-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only