missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzamidine hydriodide, 96%

Catalog No. p-7081554
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1157103 1 g
AAA1157106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1157103 Supplier Thermo Scientific Chemicals Supplier No. A1157103

CAS: 115297-57-9 | C7H8ClIN2 | 282.509 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115297-57-9
Molecular Formula C7H8ClIN2
Molecular Weight (g/mol) 282.509
MDL Number MFCD00051987
InChI Key YMEVGWRZEYGNJO-UHFFFAOYSA-N
Synonym 4-chlorobenzamidine hydroiodide, 4-chlorobenzimidamide hydroiodide, 4-chlorobenzamidine hydriodide, chlorophenamidine hydroiodide, 4-chlorobenzenecarboximidamide hydroiodide, acmc-1bu33, benzenecarboximidamide,4-chloro-,hydriodide 1:1, benzenecarboximidamide,4-chloro-, hydriodide 1:1
PubChem CID 2734221
IUPAC Name 4-chlorobenzenecarboximidamide;hydroiodide
SMILES C1=CC(=CC=C1C(=N)N)Cl.I

Specifications

Melting Point ∼235°C (decomposition)
Quantity 1 g
Beilstein 8280383
Formula Weight 282.51
Percent Purity 96%
Chemical Name or Material 4-Chlorobenzamidine hydriodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only