accessibility menu, dialog, popup

UserName

CAS RN 115297-57-9

IUPAC Name: [amino(4-chlorophenyl)methylidene]azanium
Synonyms: [amino(4-chlorophenyl)methylidene]azanium
Molecular Weight (g/mol): 155.6
Molecular Formula: C7H8ClN2
InChi Key: LFCUMSZYMJRUHD-UHFFFAOYSA-O
SMILES: NC(=[NH2+])C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results