missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzaldehyde, 98%

CAS: 104-88-1 | C7H5ClO | 140.566 g/mol

Supplier:  Thermo Scientific Chemicals A127570I

 View more versions of this product

Catalog No. AAA127570I


Only null left
Explore available promotions

Description

Description

4-Chlorobenzaldehyde is used as an intermediate for the manufacture of dyestuffs, optical brighteners, pharmaceuticals, agricultural chemicals and metal finishing products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzaldehyde is used as an intermediate for the manufacture of dyestuffs, optical brighteners, pharmaceuticals, agricultural chemicals and metal finishing products.

Solubility
Soluble in water (0.94g/L).

Notes
Air Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from air and oxidizing agent.
Specifications

Specifications

4-Chlorobenzaldehyde
45°C to 49°C
88°C (190°F)
C7H5ClO
5000 g
385858
p-chlorobenzaldehyde, benzaldehyde, 4-chloro, benzaldehyde, p-chloro, p-chlorobenzenecarboxaldehyde, 4-chloro-benzaldehyde, 4-chlorobenzenaldehyde, 4-chloro benzaldehyde, ccris 857, para-chlorobenzaldehyde, unii-e67727up9z
AVPYQKSLYISFPO-UHFFFAOYSA-N
4-chlorobenzaldehyde
7726
140.57
104-88-1
213°C to 214°C
Pungent
MFCD00003379
UN3077
Air Sensitive
Soluble in water (0.94g/L).
C1=CC(=CC=C1C=O)Cl
140.566
CHEBI:28105
98%

Chemical Identifiers

104-88-1
140.566
AVPYQKSLYISFPO-UHFFFAOYSA-N
7726
4-chlorobenzaldehyde
C7H5ClO
MFCD00003379
p-chlorobenzaldehyde, benzaldehyde, 4-chloro, benzaldehyde, p-chloro, p-chlorobenzenecarboxaldehyde, 4-chloro-benzaldehyde, 4-chlorobenzenaldehyde, 4-chloro benzaldehyde, ccris 857, para-chlorobenzaldehyde, unii-e67727up9z
CHEBI:28105
C1=CC(=CC=C1C=O)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H500

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 203-247-4

RTECSNumber : CU5076000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only