missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzaldehyde, 98%

Catalog No. AAA1275730
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1275730 250 g
AAA127570B 1000 g
AAA127570I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1275730 Supplier Thermo Scientific Chemicals Supplier No. A1275730
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104-88-1 | C7H5ClO | 140.566 g/mol

4-Chlorobenzaldehyde is used as an intermediate for the manufacture of dyestuffs, optical brighteners, pharmaceuticals, agricultural chemicals and metal finishing products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzaldehyde is used as an intermediate for the manufacture of dyestuffs, optical brighteners, pharmaceuticals, agricultural chemicals and metal finishing products.

Solubility
Soluble in water (0.94g/L).

Notes
Air Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from air and oxidizing agent.

Chemical Identifiers

CAS 104-88-1
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.566
MDL Number MFCD00003379
InChI Key AVPYQKSLYISFPO-UHFFFAOYSA-N
Synonym p-chlorobenzaldehyde, benzaldehyde, 4-chloro, benzaldehyde, p-chloro, p-chlorobenzenecarboxaldehyde, 4-chloro-benzaldehyde, 4-chlorobenzenaldehyde, 4-chloro benzaldehyde, ccris 857, para-chlorobenzaldehyde, unii-e67727up9z
PubChem CID 7726
ChEBI CHEBI:28105
IUPAC Name 4-chlorobenzaldehyde
SMILES C1=CC(=CC=C1C=O)Cl

Specifications

Melting Point 45°C to 49°C
Boiling Point 213°C to 214°C
Flash Point 88°C (190°F)
Odor Pungent
Quantity 250 g
UN Number UN3077
Beilstein 385858
Sensitivity Air Sensitive
Solubility Information Soluble in water (0.94g/L).
Formula Weight 140.57
Percent Purity 98%
Chemical Name or Material 4-Chlorobenzaldehyde
Show More Show Less
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 203-247-4
RTECSNumber CU5076000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only