Learn More
4-(Chloroacetyl)morpholine, 97+%
CAS: 1440-61-5 | C6H10ClNO2 | 163.601 g/mol
$60.74 - $206.53
Chemical Identifiers
| CAS | 1440-61-5 |
|---|---|
| Molecular Formula | C6H10ClNO2 |
| Molecular Weight (g/mol) | 163.601 |
| MDL Number | MFCD00721939 |
| InChI Key | YMQRPXBBBOXHNZ-UHFFFAOYSA-N |
| Synonym | 4-2-chloroacetyl morpholine, morpholine, 4-chloroacetyl, 4-chloroacetyl morpholine, n-chloroacetyl morpholine, 2-chloro-1-morpholin-4-yl ethan-1-one, n-chloroacetylmorpholine, chloroacetic acid morpholide, 2-chloro-1-morpholinoethanone, 2-chloro-1-morpholin-4-yl-ethanone, chloroacetic acid, morpholide |
| PubChem CID | 74040 |
| IUPAC Name | 2-chloro-1-morpholin-4-ylethanone |
| SMILES | C1COCCN1C(=O)CCl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2154803
|
Thermo Scientific Chemicals
B2154803 |
1 g |
Each for $60.74
|
|
|||||
|
AAB2154806
|
Thermo Scientific Chemicals
B2154806 |
5 g |
Each for $206.53
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1440-61-5 | |
| 163.601 | |
| YMQRPXBBBOXHNZ-UHFFFAOYSA-N | |
| 74040 | |
| C1COCCN1C(=O)CCl |
| C6H10ClNO2 | |
| MFCD00721939 | |
| 4-2-chloroacetyl morpholine, morpholine, 4-chloroacetyl, 4-chloroacetyl morpholine, n-chloroacetyl morpholine, 2-chloro-1-morpholin-4-yl ethan-1-one, n-chloroacetylmorpholine, chloroacetic acid morpholide, 2-chloro-1-morpholinoethanone, 2-chloro-1-morpholin-4-yl-ethanone, chloroacetic acid, morpholide | |
| 2-chloro-1-morpholin-4-ylethanone |
Specifications
| 1440-61-5 | |
| C6H10ClNO2 | |
| 1 g | |
| Hygroscopic | |
| YMQRPXBBBOXHNZ-UHFFFAOYSA-N | |
| 2-chloro-1-morpholin-4-ylethanone | |
| 74040 | |
| ≥97% |
| 27°C to 30°C | |
| MFCD00721939 | |
| UN3261 | |
| 4-2-chloroacetyl morpholine, morpholine, 4-chloroacetyl, 4-chloroacetyl morpholine, n-chloroacetyl morpholine, 2-chloro-1-morpholin-4-yl ethan-1-one, n-chloroacetylmorpholine, chloroacetic acid morpholide, 2-chloro-1-morpholinoethanone, 2-chloro-1-morpholin-4-yl-ethanone, chloroacetic acid, morpholide | |
| C1COCCN1C(=O)CCl | |
| 163.601 | |
| 163.61 | |
| 4-(Chloroacetyl)morpholine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 215-880-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only