missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloroacetyl)catechol 98.0+%, TCI America™

Supplier:  TCI America C015625G

Catalog No. C015625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-(Chloroacetyl)catechol
173°C
C8H7ClO3
25 g
LWTJEJCZJFZKEL-UHFFFAOYSA-N
2-chloro-1-(3,4-dihydroxyphenyl)ethanone
66834
186.59
Crystalline Powder
99-40-1
Gray-Red
MFCD00002200
2-chloro-3',4'-dihydroxyacetophenone, 2-chloro-1-3,4-dihydroxyphenyl ethanone, 4-chloroacetyl catechol, 3,4-dihydroxyphenacyl chloride, ethanone, 2-chloro-1-3,4-dihydroxyphenyl, unii-fn6949l56q, 3',4'-dihydroxy-2-chloroacetophenone, chloromethyl 3,4-dihydroxyphenyl ketone, 2-chloro-3,4-dihydroxyacetophenone, .alpha.-chloro-3,4-dihydroxyacetophenone
C1=CC(=C(C=C1C(=O)CCl)O)O
186.591
CHEBI:51844
≥98.0% (HPLC,T)

Chemical Identifiers

99-40-1
186.591
LWTJEJCZJFZKEL-UHFFFAOYSA-N
66834
2-chloro-1-(3,4-dihydroxyphenyl)ethanone
C8H7ClO3
MFCD00002200
2-chloro-3',4'-dihydroxyacetophenone, 2-chloro-1-3,4-dihydroxyphenyl ethanone, 4-chloroacetyl catechol, 3,4-dihydroxyphenacyl chloride, ethanone, 2-chloro-1-3,4-dihydroxyphenyl, unii-fn6949l56q, 3',4'-dihydroxy-2-chloroacetophenone, chloromethyl 3,4-dihydroxyphenyl ketone, 2-chloro-3,4-dihydroxyacetophenone, .alpha.-chloro-3,4-dihydroxyacetophenone
CHEBI:51844
C1=CC(=C(C=C1C(=O)CCl)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2942

TSCA : No

SDS
Documents

Documents

Promotions

Promotions