accessibility menu, dialog, popup

UserName

OHHOOCl

CAS RN 99-40-1

IUPAC Name: 2-chloro-1-(3,4-dihydroxyphenyl)ethan-1-one
Synonyms: 2-chloro-1-(3,4-dihydroxyphenyl)ethanone
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: LWTJEJCZJFZKEL-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1O)C(=O)CCl
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)