missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloroacetanilide, 98+%

Catalog No. p-7081296
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1121414 25 g
AAA1121422 100 g
AAA1121436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1121414 Supplier Thermo Scientific Chemicals Supplier No. A1121414
Only null left

CAS: 539-03-7 | C8H8ClNO | 169.608 g/mol

4'-Chloroacetanilide is used in pharmaceutical preparations, using PGC column. And also used in the synthesis of alprazolam.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Chloroacetanilide is used in pharmaceutical preparations, using PGC column. And also used in the synthesis of alprazolam.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 539-03-7
Molecular Formula C8H8ClNO
Molecular Weight (g/mol) 169.608
MDL Number MFCD00000612
InChI Key GGUOCFNAWIODMF-UHFFFAOYSA-N
Synonym 4'-chloroacetanilide, n-4-chlorophenyl acetamide, 4-chloroacetanilide, p-chloroacetanilide, n-acetyl-p-chloroaniline, acetamide, n-4-chlorophenyl, acetanilide, 4'-chloro, acetic-4-chloroanilide, n-p-chlorophenyl acetamide, n-4-chloro-phenyl-acetamide
PubChem CID 10871
ChEBI CHEBI:116915
IUPAC Name N-(4-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)Cl

Specifications

Melting Point 178°C to 180°C
Density 1.385
Boiling Point 333°C
Quantity 25 g
Beilstein 509638
Merck Index 14,2111
Solubility Information Insoluble in water.
Formula Weight 169.61
Percent Purity ≥98%
Chemical Name or Material 4'-Chloroacetanilide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-707-8
RTECSNumber AE1001000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only