missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloroacetanilide, 97%

Catalog No. AC169952500
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169952500 250 g Plastic bottle
AC169950050 5 g Glass Bottle
AC169950500 50 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC169952500 Supplier Thermo Scientific Chemicals Supplier No. 169952500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 539-03-7 | C8H8ClNO | 169.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 539-03-7
MDL Number MFCD00000612
InChI Key GGUOCFNAWIODMF-UHFFFAOYSA-N
Synonym 4'-chloroacetanilide, n-4-chlorophenyl acetamide, 4-chloroacetanilide, p-chloroacetanilide, n-acetyl-p-chloroaniline, acetamide, n-4-chlorophenyl, acetanilide, 4'-chloro, acetic-4-chloroanilide, n-p-chlorophenyl acetamide, n-4-chloro-phenyl-acetamide
PubChem CID 10871
ChEBI CHEBI:116915
IUPAC Name N-(4-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 177°C to 181°C
Color Beige-Gray or White
Boiling Point 333°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Molecular Formula C8H8ClNO
Linear Formula CH3CONHC6H4Cl
Quantity 250 g
Beilstein 12, 611
Merck Index 14, 2111
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,ether and carbon disulfide,slightly soluble in ccl4 and benzene
Molecular Weight (g/mol) 169.61
Formula Weight 169.61
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloroacetanilide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-707-8

RUO – Research Use Only