missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-o-xylene 98.0+%, TCI America™

Catalog No. C035625G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C035625G 25 g
C0356100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C035625G Supplier TCI America Supplier No. C035625G

Chemical Identifiers

CAS 615-60-1
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000596
InChI Key HNQLMBJUMVLFCF-UHFFFAOYSA-N
Synonym 4-chloro-o-xylene, 3,4-dimethylchlorobenzene, 1-chloro-3,4-dimethylbenzene, benzene, 4-chloro-1,2-dimethyl, o-xylene, 4-chloro, 4-chloro-ortho-xylene, 4-chloro-1,2-xylene, unii-qo24waw04q, qo24waw04q, pubchem3641
PubChem CID 32886
IUPAC Name 4-chloro-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)Cl)C

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 192°C
Formula Weight 140.61
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Chloro-o-xylene