missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-methylpyridine-2-carboxamide, 97%

Catalog No. p-7033424
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50535MD 250 mg
AAH5053503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50535MD Supplier Thermo Scientific Chemicals Supplier No. H50535MD

CAS: 220000-87-3 | C7H7ClN2O | 170.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 220000-87-3
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.60
MDL Number MFCD02185921
InChI Key BGVBBMZMEKXUTR-UHFFFAOYSA-N
Synonym 4-chloro-n-methylpicolinamide, n-methyl-4-chloropyridine-2-carboxamide, 4-chloro-n-methyl-2-pyridinecarboxamide, n-methyl 4-chloropicolinamide, 2-pyridinecarboxamide, 4-chloro-n-methyl, 4-chloropyridine-2-carboxylic acid methylamide, 4-chloro-n-methyl-pyridine-2-carboxamide, n-methyl-4-chloro-2-pyridinamide, 4-chloro 2-pyridyl-n-methylcarboxamide, 4-chloranyl-∼ n-methyl-pyridine-2-carboxamide
PubChem CID 1476814
IUPAC Name 4-chloro-N-methylpyridine-2-carboxamide
SMILES CNC(=O)C1=NC=CC(Cl)=C1

Specifications

Melting Point 39°C to 43°C
Quantity 250 mg
Formula Weight 170.6
Percent Purity 97%
Chemical Name or Material 4-Chloro-N-methylpyridine-2-carboxamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only