accessibility menu, dialog, popup

UserName

CH3HNONCl

CAS RN 220000-87-3

IUPAC Name: 4-chloro-N-methylpyridine-2-carboxamide
Synonyms: 4-chloro-N-methylpyridine-2-carboxamide
Molecular Weight (g/mol): 170.6
Molecular Formula: C7H7ClN2O
InChi Key: BGVBBMZMEKXUTR-UHFFFAOYSA-N
SMILES: CNC(=O)C1=NC=CC(Cl)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)