missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-methyl-2-pyridinecarboxamide 98.0+%, TCI America™

Catalog No. C27935G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27935G 5 g
C27931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27935G Supplier TCI America Supplier No. C27935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 220000-87-3 | C7H7ClN2O | 170.60 g/mol

Chemical Identifiers

CAS 220000-87-3
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.60
MDL Number MFCD02185921
InChI Key BGVBBMZMEKXUTR-UHFFFAOYSA-N
Synonym 4-chloro-n-methylpicolinamide, n-methyl-4-chloropyridine-2-carboxamide, 4-chloro-n-methyl-2-pyridinecarboxamide, n-methyl 4-chloropicolinamide, 2-pyridinecarboxamide, 4-chloro-n-methyl, 4-chloropyridine-2-carboxylic acid methylamide, 4-chloro-n-methyl-pyridine-2-carboxamide, n-methyl-4-chloro-2-pyridinamide, 4-chloro 2-pyridyl-n-methylcarboxamide, 4-chloranyl-∼ n-methyl-pyridine-2-carboxamide
PubChem CID 1476814
IUPAC Name 4-chloro-N-methylpyridine-2-carboxamide
SMILES CNC(=O)C1=NC=CC(Cl)=C1

Specifications

Melting Point 47°C
Color Yellow
Quantity 5 g
Formula Weight 170.60
Percent Purity ≥98.0% (HPLC,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-N-methyl-2-pyridinecarboxamide