missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-(4-methoxyphenyl)benzamide, 97%

Catalog No. p-7036969
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55469MD 250 mg
AAH5546903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH55469MD Supplier Thermo Scientific Chemicals Supplier No. H55469MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4018-82-0 | C14H12ClNO2 | 261.705 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4018-82-0
Molecular Formula C14H12ClNO2
Molecular Weight (g/mol) 261.705
MDL Number MFCD00508611
InChI Key UVGPQLMHIRNFEE-UHFFFAOYSA-N
Synonym 4-chloro-n-4-methoxyphenyl benzamide, n-p-chlorobenzoyl-p-anisidine, 4-chloro-n-4-methoxy-phenyl-benzamide, benzamide, 4-chloro-n-4-methoxyphenyl, cambridge id 5190257, n-4-methoxyphenyl-4-chlorobenzamide, 4-chloro-n-4-methoxyphenyl-benzamide, 4-chloro-n-4-methoxyphenyl benzamide #, 4-chlorobenzenamide, n-4-methoxyphenyl
PubChem CID 221848
IUPAC Name 4-chloro-N-(4-methoxyphenyl)benzamide
SMILES COC1=CC=C(C=C1)NC(=O)C2=CC=C(C=C2)Cl

Specifications

Quantity 250 mg
Formula Weight 261.71
Percent Purity 97%
Chemical Name or Material 4-Chloro-N-(4-methoxyphenyl)benzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only