missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide 98.0+%, TCI America™
$88.56 - $442.79
Chemical Identifiers
| CAS | 41859-57-8 |
|---|---|
| Molecular Formula | C15H14ClNO2 |
| Molecular Weight (g/mol) | 275.73 |
| MDL Number | MFCD00218513 |
| InChI Key | ZTLWJYCDAXUIBK-UHFFFAOYSA-N |
| Synonym | n-4-chlorobenzoyl-tyramine, 4-chloro-n-4-hydroxyphenethyl benzamide, 4-chloro-n-2-4-hydroxyphenyl ethyl benzamide, n-4-chlorobenzoyl tyramine, benzamide, p-chloro-n-2-p-hydroxyphenyl ethyl, p-chloro-n-2-p-hydroxyphenyl ethyl benzamide, benzamide, 4-chloro-n-2-4-hydroxyphenyl ethyl, bez-da |
| PubChem CID | 198234 |
| IUPAC Name | 4-chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide |
| SMILES | OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1 |
Chemical Identifiers
| 41859-57-8 | |
| 275.73 | |
| ZTLWJYCDAXUIBK-UHFFFAOYSA-N | |
| 198234 | |
| OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1 |
| C15H14ClNO2 | |
| MFCD00218513 | |
| n-4-chlorobenzoyl-tyramine, 4-chloro-n-4-hydroxyphenethyl benzamide, 4-chloro-n-2-4-hydroxyphenyl ethyl benzamide, n-4-chlorobenzoyl tyramine, benzamide, p-chloro-n-2-p-hydroxyphenyl ethyl, p-chloro-n-2-p-hydroxyphenyl ethyl benzamide, benzamide, 4-chloro-n-2-4-hydroxyphenyl ethyl, bez-da | |
| 4-chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide |
Specifications
| 41859-57-8 | |
| C15H14ClNO2 | |
| 1 g | |
| ZTLWJYCDAXUIBK-UHFFFAOYSA-N | |
| 4-chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide | |
| 198234 | |
| ≥98.0% (HPLC,N) | |
| 4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide |
| 174°C | |
| MFCD00218513 | |
| n-4-chlorobenzoyl-tyramine, 4-chloro-n-4-hydroxyphenethyl benzamide, 4-chloro-n-2-4-hydroxyphenyl ethyl benzamide, n-4-chlorobenzoyl tyramine, benzamide, p-chloro-n-2-p-hydroxyphenyl ethyl, p-chloro-n-2-p-hydroxyphenyl ethyl benzamide, benzamide, 4-chloro-n-2-4-hydroxyphenyl ethyl, bez-da | |
| OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1 | |
| 275.73 | |
| 275.73 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : CV2450481
TSCA : No