missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-m-xylene 98.0+%, TCI America™
$104.38 - $307.57
Chemical Identifiers
| CAS | 95-66-9 |
|---|---|
| Molecular Formula | C8H9Cl |
| Molecular Weight (g/mol) | 140.61 |
| MDL Number | MFCD00060644 |
| InChI Key | UIEVCEQLNUHDIF-UHFFFAOYSA-N |
| Synonym | 4-chloro-m-xylene, m-xylene, 4-chloro, 2,4-dimethylchlorobenzene, benzene, 1-chloro-2,4-dimethyl, mxylene4chloro, 1-chloro-2,4-dimethyl-benzene, 4-chlor-m-xylol, pubchem3639, acmc-209rzq, 4-chloro-meta-xylene |
| PubChem CID | 523153 |
| IUPAC Name | 1-chloro-2,4-dimethylbenzene |
| SMILES | CC1=CC(=C(C=C1)Cl)C |
Chemical Identifiers
| 95-66-9 | |
| 140.61 | |
| UIEVCEQLNUHDIF-UHFFFAOYSA-N | |
| 523153 | |
| CC1=CC(=C(C=C1)Cl)C |
| C8H9Cl | |
| MFCD00060644 | |
| 4-chloro-m-xylene, m-xylene, 4-chloro, 2,4-dimethylchlorobenzene, benzene, 1-chloro-2,4-dimethyl, mxylene4chloro, 1-chloro-2,4-dimethyl-benzene, 4-chlor-m-xylol, pubchem3639, acmc-209rzq, 4-chloro-meta-xylene | |
| 1-chloro-2,4-dimethylbenzene |
Specifications
| -32°C | |
| Yellow | |
| 5 g | |
| C8H9Cl | |
| 4-chloro-m-xylene, m-xylene, 4-chloro, 2,4-dimethylchlorobenzene, benzene, 1-chloro-2,4-dimethyl, mxylene4chloro, 1-chloro-2,4-dimethyl-benzene, 4-chlor-m-xylol, pubchem3639, acmc-209rzq, 4-chloro-meta-xylene | |
| CC1=CC(=C(C=C1)Cl)C | |
| 140.61 | |
| 140.61 | |
| 4-Chloro-m-xylene |
| 95-66-9 | |
| 188°C | |
| Clear Liquid at 20°C | |
| MFCD00060644 | |
| UIEVCEQLNUHDIF-UHFFFAOYSA-N | |
| 1-chloro-2,4-dimethylbenzene | |
| 523153 | |
| ≥98.0% (GC) |
Safety and Handling
TSCA : No