missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-m-xylene 98.0+%, TCI America™

Catalog No. C21655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21655G 5 g
C216525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21655G Supplier TCI America Supplier No. C21655G

Chemical Identifiers

CAS 95-66-9
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00060644
InChI Key UIEVCEQLNUHDIF-UHFFFAOYSA-N
Synonym 4-chloro-m-xylene, m-xylene, 4-chloro, 2,4-dimethylchlorobenzene, benzene, 1-chloro-2,4-dimethyl, mxylene4chloro, 1-chloro-2,4-dimethyl-benzene, 4-chlor-m-xylol, pubchem3639, acmc-209rzq, 4-chloro-meta-xylene
PubChem CID 523153
IUPAC Name 1-chloro-2,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)Cl)C

Specifications

Melting Point -32°C
Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 188°C
Formula Weight 140.61
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Chloro-m-xylene