missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-m-cresol 99.0+%, TCI America™

Catalog No. p-7124249
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C015025G 25 g
C0150500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C015025G Supplier TCI America Supplier No. C015025G
Only null left

Chemical Identifiers

CAS 59-50-7
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00002323
InChI Key CFKMVGJGLGKFKI-UHFFFAOYSA-N
Synonym chlorocresol, 4-chloro-m-cresol, baktol, parol, p-chloro-m-cresol, ottafact, phenol, 4-chloro-3-methyl, 4-chloro-3-cresol, candaseptic, baktolan
PubChem CID 1732
ChEBI CHEBI:34395
IUPAC Name 4-chloro-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)Cl

Specifications

Melting Point 66°C
Color White
Boiling Point 235°C
Quantity 25 g
UN Number 3437
Formula Weight 142.58
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-m-cresol
EINECSNumber (3)-0900
RTECSNumber GO7100000
TSCA Yes