missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-L-phenylalanine, 97%

Catalog No. p-7055082
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63573MD 250 mg
AAH6357303 1 g
AAH6357306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63573MD Supplier Thermo Scientific Chemicals Supplier No. H63573MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14173-39-8 | C9H10ClNO2 | 199.63 g/mol

4-Chloro-L-phenylalanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-L-phenylalanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Soluble in acetic acid. Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 14173-39-8
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD00063065,MFCD00079675
InChI Key NIGWMJHCCYYCSF-UHFFFAOYNA-N
Synonym 4-chloro-l-phenylalanine, l-4-chlorophenylalanine, h-phe 4-cl-oh, s-2-amino-3-4-chlorophenyl propanoic acid, l-p-chlorophenylalanine, 2s-2-amino-3-4-chlorophenyl propanoic acid, fenclonine, s, l-pcpa, unii-snm151oe2c, l-phe 4-cl-oh
PubChem CID 736190
SMILES NC(CC1=CC=C(Cl)C=C1)C(O)=O

Specifications

Melting Point 235°C to 238°C
UN Number UN2811
Beilstein 2416150
Solubility Information Soluble in acetic acid. Slightly soluble in water.
IUPAC Name (2S)-2-amino-3-(4-chlorophenyl)propanoic acid
Quantity 250 mg
Formula Weight 199.63
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-L-phenylalanine
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 238-023-5
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only