CAS RN 14173-39-8
IUPAC Name:
2-amino-3-(4-chlorophenyl)propanoic acid
Synonyms:
fenclonine fenclonine hydrochloride fenclonine (L)- fenclonina P-chlorophenylalanine,DL 4-chloro-3-phenylalanine p-clorophenylalanine (D)-fenclonine fenclonine, (D)- fenchlonine
Molecular Weight (g/mol):
199.63
Molecular Formula:
C9H10ClNO2
InChi Key:
NIGWMJHCCYYCSF-UHFFFAOYNA-N
SMILES:
NC(CC1=CC=C(Cl)C=C1)C(O)=O
1
–
1
of
1
results
1
Promotions Available
4-Chloro-L-phenylalanine, 97%
4-Chloro-L-phenylalanine, 97%
CAS: 14173-39-8 Molecular Formula: C9H10ClNO2 Molecular Weight (g/mol): 199.63 MDL Number: MFCD00063065,MFCD00079675 InChI Key: NIGWMJHCCYYCSF-UHFFFAOYNA-N Synonym: 4-chloro-l-phenylalanine,l-4-chlorophenylalanine,h-phe 4-cl-oh,s-2-amino-3-4-chlorophenyl propanoic acid,l-p-chlorophenylalanine,2s-2-amino-3-4-chlorophenyl propanoic acid,fenclonine, s,l-pcpa,unii-snm151oe2c,l-phe 4-cl-oh PubChem CID: 736190 SMILES: NC(CC1=CC=C(Cl)C=C1)C(O)=O