missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-DL-phenylglycine, 98%

Catalog No. p-7053434
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5992114 25 g
AAH5992122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5992114 Supplier Thermo Scientific Chemicals Supplier No. H5992114

CAS: 6212-33-5 | C8H8ClNO2 | 185.607 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 0.1mol/L NaOH (almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 6212-33-5
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD00049328
InChI Key QGJGBYXRJVIYGA-UHFFFAOYSA-N
Synonym 2-amino-2-4-chlorophenyl acetic acid, amino 4-chlorophenyl acetic acid, amino-4-chloro-phenyl-acetic acid, dl-2-4-chlorophenyl glycine, 2-4-chlorophenyl glycine, +/--4-chlorophenylglycine, h-dl-phg 4-cl-oh, 4-chloro-dl-phenylglycine, dl-4-chlorophenyl glycine, 4-chlorophenyl glycine
PubChem CID 587821
IUPAC Name 2-amino-2-(4-chlorophenyl)acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)N)Cl

Specifications

Color White
Solubility Information Soluble in 0.1mol/L NaOH (almost transparency).
Quantity 25 g
Formula Weight 185.61
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Chloro-DL-phenylglycine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only