missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-Chloro-D-phenylalanine, 95%

CAS: 14091-08-8 | C9H10ClNO2 | 199.63 g/mol

Supplier:  Thermo Scientific Chemicals H5198206

Quantity 5 g

 View more versions of this product

Catalog No. AAH5198206


Only null left

Description

Description

It is applied in the synthesis and placental binding potencies of photosensitive analogues of luteinizing hormone releasing hormone (lhrh) with agonistic and antagonistic structures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in the synthesis and placental binding potencies of photosensitive analogues of luteinizing hormone releasing hormone (lhrh) with agonistic and antagonistic structures.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

14091-08-8
199.63
NIGWMJHCCYYCSF-UHFFFAOYNA-N
667429
NC(CC1=CC=C(Cl)C=C1)C(O)=O
C9H10ClNO2
MFCD00079675
4-chloro-d-phenylalanine, d-4-chlorophenylalanine, r-2-amino-3-4-chlorophenyl propanoic acid, d-phenylalanine, 4-chloro, 2r-2-amino-3-4-chlorophenyl propanoic acid, h-d-phe 4-cl-oh, fenclonine, r, d-p-chlorophenylalanine, unii-rss1jq7mes, rss1jq7mes
(2R)-2-amino-3-(4-chlorophenyl)propanoic acid

Specifications

4-Chloro-D-phenylalanine
14091-08-8
MFCD00079675
2416151
Slightly soluble in water.
NC(CC1=CC=C(Cl)C=C1)C(O)=O
199.63
667429
95%
260°C
C9H10ClNO2
UN2811
4-chloro-d-phenylalanine, d-4-chlorophenylalanine, r-2-amino-3-4-chlorophenyl propanoic acid, d-phenylalanine, 4-chloro, 2r-2-amino-3-4-chlorophenyl propanoic acid, h-d-phe 4-cl-oh, fenclonine, r, d-p-chlorophenylalanine, unii-rss1jq7mes, rss1jq7mes
NIGWMJHCCYYCSF-UHFFFAOYNA-N
(2R)-2-amino-3-(4-chlorophenyl)propanoic acid
5 g
199.64
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280g-P301+P310-P302+P352-P330-P333+P313-P363-P501c

H301-H317

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions