missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-Chloro-D-phenylalanine, 95%

Catalog No. p-7034311
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51982MD 250 mg
AAH5198203 1 g
AAH5198206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH51982MD Supplier Thermo Scientific Chemicals Supplier No. H51982MD

CAS: 14091-08-8 | C9H10ClNO2 | 199.63 g/mol

It is applied in the synthesis and placental binding potencies of photosensitive analogues of luteinizing hormone releasing hormone (lhrh) with agonistic and antagonistic structures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in the synthesis and placental binding potencies of photosensitive analogues of luteinizing hormone releasing hormone (lhrh) with agonistic and antagonistic structures.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 14091-08-8
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD00079675
InChI Key NIGWMJHCCYYCSF-UHFFFAOYNA-N
Synonym 4-chloro-d-phenylalanine, d-4-chlorophenylalanine, r-2-amino-3-4-chlorophenyl propanoic acid, d-phenylalanine, 4-chloro, 2r-2-amino-3-4-chlorophenyl propanoic acid, h-d-phe 4-cl-oh, fenclonine, r, d-p-chlorophenylalanine, unii-rss1jq7mes, rss1jq7mes
PubChem CID 667429
IUPAC Name (2R)-2-amino-3-(4-chlorophenyl)propanoic acid
SMILES NC(CC1=CC=C(Cl)C=C1)C(O)=O

Specifications

Melting Point 260°C
UN Number UN2811
Beilstein 2416151
Solubility Information Slightly soluble in water.
Quantity 250 mg
Formula Weight 199.64
Percent Purity 95%
Chemical Name or Material 4-Chloro-D-phenylalanine
Hazard Category H301-H317
Hazard Statement GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P310-P302+P352-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only