missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-alpha-methylphenylacetic acid, 97%

CAS: 938-95-4 | C9H9ClO2 | 184.619 g/mol

Supplier:  Thermo Scientific Chemicals L0602014

 View more versions of this product

Catalog No. AAL0602014

Only null left
Explore available promotions

Description

Description

4-Chloro-alpha-methylphenylacetic acid is used to produce a-(4-Chlor-3-nitrophenyl)propionsaure. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-alpha-methylphenylacetic acid is used to produce a-(4-Chlor-3-nitrophenyl)propionsaure. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications

Specifications

4-Chloro-alpha-methylphenylacetic acid
55°C to 59°C
C9H9ClO2
25 g
2-4-chlorophenyl propanoic acid, 4-chloro-alpha-methylphenylacetic acid, dl-2-4-chlorophenyl propan-oic acid, dl-p-chloro-.alpha.-methylphenylacetic acid, benzeneacetic acid, 4-chloro-.alpha.-methyl, dl-2-4-chlorophenyl propanoic acid, 2-4-chloro-phenyl-propionic acid, dl-4-chloro-alpha-methylphenylacetic acid, ?-p-chlorohydratropic acid, 4-chloro methylphenylacetic acid
YOZILQVNIWNPFP-UHFFFAOYSA-N
2-(4-chlorophenyl)propanoic acid
102525
97%
938-95-4
121°C to 126°C (0.2 mmHg)
MFCD00044670
2328710
Soluble in water.
CC(C1=CC=C(C=C1)Cl)C(=O)O
184.619
184.62

Chemical Identifiers

938-95-4
184.619
YOZILQVNIWNPFP-UHFFFAOYSA-N
102525
CC(C1=CC=C(C=C1)Cl)C(=O)O
C9H9ClO2
MFCD00044670
2-4-chlorophenyl propanoic acid, 4-chloro-alpha-methylphenylacetic acid, dl-2-4-chlorophenyl propan-oic acid, dl-p-chloro-.alpha.-methylphenylacetic acid, benzeneacetic acid, 4-chloro-.alpha.-methyl, dl-2-4-chlorophenyl propanoic acid, 2-4-chloro-phenyl-propionic acid, dl-4-chloro-alpha-methylphenylacetic acid, ?-p-chlorohydratropic acid, 4-chloro methylphenylacetic acid
2-(4-chlorophenyl)propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P272-P280-P301+P310-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c

H301-H317-H319

EINECSNumber : 213-351-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only