missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-alpha-methylphenylacetic acid, 97%

Catalog No. p-7044359
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0602006 5 g
AAL0602014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0602006 Supplier Thermo Scientific Chemicals Supplier No. L0602006
Only null left

CAS: 938-95-4 | C9H9ClO2 | 184.619 g/mol

4-Chloro-alpha-methylphenylacetic acid is used to produce a-(4-Chlor-3-nitrophenyl)propionsaure. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-alpha-methylphenylacetic acid is used to produce a-(4-Chlor-3-nitrophenyl)propionsaure. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Chemical Identifiers

CAS 938-95-4
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00044670
InChI Key YOZILQVNIWNPFP-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl propanoic acid, 4-chloro-alpha-methylphenylacetic acid, dl-2-4-chlorophenyl propan-oic acid, dl-p-chloro-.alpha.-methylphenylacetic acid, benzeneacetic acid, 4-chloro-.alpha.-methyl, dl-2-4-chlorophenyl propanoic acid, 2-4-chloro-phenyl-propionic acid, dl-4-chloro-alpha-methylphenylacetic acid, ?-p-chlorohydratropic acid, 4-chloro methylphenylacetic acid
PubChem CID 102525
IUPAC Name 2-(4-chlorophenyl)propanoic acid
SMILES CC(C1=CC=C(C=C1)Cl)C(=O)O

Specifications

Melting Point 55°C to 59°C
Boiling Point 121°C to 126°C (0.2 mmHg)
Quantity 5 g
Beilstein 2328710
Solubility Information Soluble in water.
Formula Weight 184.62
Percent Purity 97%
Chemical Name or Material 4-Chloro-alpha-methylphenylacetic acid
Hazard Category H301-H317-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P310-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c
EINECSNumber 213-351-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only