missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-7-methyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine, 96%

Catalog No. AAH3343903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3343903 1 g
AAH3343906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3343903 Supplier Thermo Scientific Chemicals Supplier No. H3343903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 137438-23-4 | C11H11ClN2S | 238.733 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 137438-23-4
Molecular Formula C11H11ClN2S
Molecular Weight (g/mol) 238.733
MDL Number MFCD02333821
InChI Key ZSDWARZBRRSRLT-UHFFFAOYSA-N
Synonym 4-chloro-7-methyl-5,6,7,8-tetrahydro 1 benzothieno 2,3-d pyrimidine, 4-chloro-7-methyl-5,6,7,8-tetrahydrobenzo 4,5 thieno 2,3-d pyrimidine, 4-chloro-7-methyl-5,6,7,8-tetrahydrobenzo b thiopheno 2,3-d pyrimidine, 3-chloro-11-methyl-8-thia-4,6-diazatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6-tetraene, rs-4-chloro-7-methyl-5,6,7,8-tetrahydro 1 benzothieno 2,3-d pyrimidine, 4-chloro-7-methyl-5,6,7,8-tetrahydro-1 benzothiolo 2,3-d pyrimidine, 4-chloro-7-methyl-5,6,7,8-tetrahydrobenzo b thieno 2,3-d pyrimidine, 3-chloro-11-methyl-8-thia-4,6-diazatricyclo 7.4.0.0 2 , trideca-1 9 ,2,4,6-tetraene, 3-chloro-11-methyl-8-thia-4,6-diazatricyclo 7.4.0.0?,? trideca-1 9 ,2,4,6-tetraene, 3-chloro-11-methyl-8-thia-4,6-diazatricyclo 7.4.0.0^ 2,7 trideca-1 9 ,2 7 ,3,5-tetraene
PubChem CID 2790841
IUPAC Name 4-chloro-7-methyl-5,6,7,8-tetrahydro-[1]benzothiolo[2,3-d]pyrimidine
SMILES CC1CCC2=C(C1)SC3=C2C(=NC=N3)Cl

Specifications

Melting Point 109°C to 113°C
Quantity 1 g
Formula Weight 238.73
Percent Purity 96%
Chemical Name or Material 4-Chloro-7-methyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only