missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-7-chlorosulfonyl-2,1,3-benzoxadiazole 97.0+%, TCI America™

Catalog No. p-7124951
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13941G 1 g
C13945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13941G Supplier TCI America Supplier No. C13941G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142246-48-8
Molecular Formula C6H2Cl2N2O3S
Molecular Weight (g/mol) 253.053
MDL Number MFCD00191405
InChI Key BPPRLMZEVZSIIJ-UHFFFAOYSA-N
Synonym 4-Chloro-2,1,3-benzoxadiazole-7-sulfonyl Chloride, 4-Chloro-7-chlorosulfonylbenzofurazan
PubChem CID 612515
IUPAC Name 4-chloro-2,1,3-benzoxadiazole-7-sulfonyl chloride
SMILES C1=C(C2=NON=C2C(=C1)Cl)S(=O)(=O)Cl

Specifications

Melting Point 87°C
Quantity 1 g
UN Number 3261
Formula Weight 253.05
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-7-chlorosulfonyl-2,1,3-benzoxadiazole [Bifunctional Fluorescent Reagent]