missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6-nitro-m-cresol 98.0+%, TCI America™

Catalog No. C16845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16845G 5 g
C168425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16845G Supplier TCI America Supplier No. C16845G
Only null left

Chemical Identifiers

CAS 7147-89-9
Molecular Formula C7H6ClNO3
Molecular Weight (g/mol) 187.58
MDL Number MFCD00007114
InChI Key JBMGJOKJUYGIJH-UHFFFAOYSA-N
PubChem CID 23532
IUPAC Name 4-chloro-5-methyl-2-nitrophenol
SMILES CC1=CC(O)=C(C=C1Cl)[N+]([O-])=O

Specifications

Melting Point 134°C
Quantity 5 g
Synonym 4-chloro-6-nitro-m-cresol, 4-chloro-3-methyl-6-nitrophenol, phenol, 4-chloro-5-methyl-2-nitro, m-cresol, 4-chloro-6-nitro, acmc-1bjod, 4-chloro-6-nitro-meta-cresol, 4-chloro 3-methyl 6-nitrophenol, 4-chloro-5-methyl-2-nitro-phenol, #, phenol,4-chloro-5-methyl-2-nitro
Formula Weight 187.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-6-nitro-m-cresol
EINECSNumber (3)-0826&(3)-0938
TSCA Yes