missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-6-fluoroquinoline 98.0+%, TCI America™
$236.73 - $814.59
Chemical Identifiers
| CAS | 391-77-5 |
|---|---|
| Molecular Formula | C9H5ClFN |
| Molecular Weight (g/mol) | 181.59 |
| MDL Number | MFCD00278783 |
| InChI Key | CKTQPWIDUQGUGG-UHFFFAOYSA-N |
| Synonym | 4-chloro-6-fluoro-quinoline, quinoline, 4-chloro-6-fluoro, pubchem5845, 6-fluoro-4-chloroquinoline, ksc493m4t, 4-chloranyl-6-fluoranyl-quinoline, 4-chloro-6-fluoroquinoline |
| PubChem CID | 2736586 |
| IUPAC Name | 4-chloro-6-fluoroquinoline |
| SMILES | FC1=CC=C2N=CC=C(Cl)C2=C1 |
Chemical Identifiers
| 391-77-5 | |
| 181.59 | |
| CKTQPWIDUQGUGG-UHFFFAOYSA-N | |
| 2736586 | |
| FC1=CC=C2N=CC=C(Cl)C2=C1 |
| C9H5ClFN | |
| MFCD00278783 | |
| 4-chloro-6-fluoro-quinoline, quinoline, 4-chloro-6-fluoro, pubchem5845, 6-fluoro-4-chloroquinoline, ksc493m4t, 4-chloranyl-6-fluoranyl-quinoline, 4-chloro-6-fluoroquinoline | |
| 4-chloro-6-fluoroquinoline |
Specifications
| 391-77-5 | |
| Yellow | |
| MFCD00278783 | |
| 4-chloro-6-fluoro-quinoline, quinoline, 4-chloro-6-fluoro, pubchem5845, 6-fluoro-4-chloroquinoline, ksc493m4t, 4-chloranyl-6-fluoranyl-quinoline, 4-chloro-6-fluoroquinoline | |
| FC1=CC=C2N=CC=C(Cl)C2=C1 | |
| 181.59 | |
| 181.59 | |
| Crystalline Powder |
| 77°C | |
| C9H5ClFN | |
| 1 g | |
| CKTQPWIDUQGUGG-UHFFFAOYSA-N | |
| 4-chloro-6-fluoroquinoline | |
| 2736586 | |
| ≥98.0% (GC) | |
| 4-Chloro-6-fluoroquinoline |
Safety and Handling
TSCA : No