CAS RN 391-77-5
IUPAC Name:
4-chloro-6-fluoroquinoline
Synonyms:
4-chloro-6-fluoroquinoline
Molecular Weight (g/mol):
181.59
Molecular Formula:
C9H5ClFN
InChi Key:
CKTQPWIDUQGUGG-UHFFFAOYSA-N
SMILES:
FC1=CC=C2N=CC=C(Cl)C2=C1
1
–
1
of
1
results
1
4-Chloro-6-fluoroquinoline 98.0+%, TCI America™
CAS: 391-77-5 Molecular Formula: C9H5ClFN Molecular Weight (g/mol): 181.59 MDL Number: MFCD00278783 InChI Key: CKTQPWIDUQGUGG-UHFFFAOYSA-N Synonym: 4-chloro-6-fluoro-quinoline,quinoline, 4-chloro-6-fluoro,pubchem5845,6-fluoro-4-chloroquinoline,ksc493m4t,4-chloranyl-6-fluoranyl-quinoline,4-chloro-6-fluoroquinoline PubChem CID: 2736586 IUPAC Name: 4-chloro-6-fluoroquinoline SMILES: FC1=CC=C2N=CC=C(Cl)C2=C1