missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-6-fluoroquinoline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 77°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 181.59 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Chloro-6-fluoroquinoline |
Safety and Handling
| TSCA | No |