missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6-fluoroquinoline 98.0+%, TCI America™

Catalog No. C29891G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29891G 1 g
C29895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29891G Supplier TCI America Supplier No. C29891G
Only null left

Chemical Identifiers

CAS 391-77-5
Molecular Formula C9H5ClFN
Molecular Weight (g/mol) 181.59
MDL Number MFCD00278783
InChI Key CKTQPWIDUQGUGG-UHFFFAOYSA-N
Synonym 4-chloro-6-fluoro-quinoline, quinoline, 4-chloro-6-fluoro, pubchem5845, 6-fluoro-4-chloroquinoline, ksc493m4t, 4-chloranyl-6-fluoranyl-quinoline, 4-chloro-6-fluoroquinoline
PubChem CID 2736586
IUPAC Name 4-chloro-6-fluoroquinoline
SMILES FC1=CC=C2N=CC=C(Cl)C2=C1

Specifications

Melting Point 77°C
Color Yellow
Quantity 1 g
Formula Weight 181.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-6-fluoroquinoline