missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6,7-dimethoxyquinazoline 98.0+%, TCI America™

Catalog No. p-7125536
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25745G 5 g
C257425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25745G Supplier TCI America Supplier No. C25745G
Only null left

Chemical Identifiers

CAS 13790-39-1
Molecular Formula C10H9ClN2O2
Molecular Weight (g/mol) 224.64
MDL Number MFCD01570172
InChI Key LLLHRNQLGUOJHP-UHFFFAOYSA-N
Synonym 4-chloro-6,7-dimethoxy-quinazoline, quinazoline, 4-chloro-6,7-dimethoxy, 4-cholro-6,7-dimethoxyquinazoline, 6,7-dimethoxy-4-chloroquinazoline, zlchem 87, pubchem9672, acmc-1brd3, 4-chloro-6,7dimethoxyquinazoline, 4-chloro6,7-dimethoxyquinazoline, 4chloro-6,7-dimethoxyquinazoline
PubChem CID 2769364
IUPAC Name 4-chloro-6,7-dimethoxyquinazoline
SMILES COC1=C(OC)C=C2C(Cl)=NC=NC2=C1

Specifications

Melting Point 185°C
Color Yellow
Quantity 5 g
Formula Weight 224.64
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-6,7-dimethoxyquinazoline