accessibility menu, dialog, popup

UserName

CAS RN 13790-39-1

IUPAC Name: 4-chloro-6,7-dimethoxyquinazoline
Synonyms: 4-chloro-6,7-dimethoxyquinazoline
Molecular Weight (g/mol): 224.64
Molecular Formula: C10H9ClN2O2
InChi Key: LLLHRNQLGUOJHP-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C(Cl)=NC=NC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)