missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6,7-dimethoxyquinazoline, 98%

Catalog No. p-7030399
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2840506 5 g
AAH2840514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2840506 Supplier Thermo Scientific Chemicals Supplier No. H2840506

CAS: 13790-39-1 | C10H9ClN2O2 | 224.64 g/mol

A useful synthetic intermediate in the preparation of epidermal growth factors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful synthetic intermediate in the preparation of epidermal growth factors.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 13790-39-1
Molecular Formula C10H9ClN2O2
Molecular Weight (g/mol) 224.64
MDL Number MFCD01570172
InChI Key LLLHRNQLGUOJHP-UHFFFAOYSA-N
Synonym 4-chloro-6,7-dimethoxy-quinazoline, quinazoline, 4-chloro-6,7-dimethoxy, 4-cholro-6,7-dimethoxyquinazoline, 6,7-dimethoxy-4-chloroquinazoline, zlchem 87, pubchem9672, acmc-1brd3, 4-chloro-6,7dimethoxyquinazoline, 4-chloro6,7-dimethoxyquinazoline, 4chloro-6,7-dimethoxyquinazoline
PubChem CID 2769364
IUPAC Name 4-chloro-6,7-dimethoxyquinazoline
SMILES COC1=C(OC)C=C2C(Cl)=NC=NC2=C1

Specifications

Melting Point 184°C to 187°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 224.65
Percent Purity 98%
Chemical Name or Material 4-Chloro-6,7-dimethoxyquinazoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only