missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-5'-phenyl-1,1':3',1″-terphenyl 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 143°C |
| Color | White |
| Quantity | 200 mg |
| Formula Weight | 340.85 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Chloro-5′-phenyl-1,1′:3′,1′′-terphenyl |
Safety and Handling
| TSCA | No |