missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5-fluoro-o-phenylenediamine, 98%

CAS: 139512-70-2 | C6H6ClFN2 | 160.58 g/mol

$100.45 - $379.53

Chemical Identifiers

CAS 139512-70-2
Molecular Formula C6H6ClFN2
Molecular Weight (g/mol) 160.58
MDL Number MFCD00042485
InChI Key BSMPRJISGCTCDC-UHFFFAOYSA-N
PubChem CID 735773
IUPAC Name 4-chloro-5-fluorobenzene-1,2-diamine
SMILES NC1=CC(F)=C(Cl)C=C1N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1395303
View Documents
Thermo Scientific Chemicals
A1395303
1 g
Each for $100.45
Only null left
 
AAA1395306
View Documents
Thermo Scientific Chemicals
A1395306
5 g
Each for $379.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

139512-70-2
160.58
BSMPRJISGCTCDC-UHFFFAOYSA-N
4-chloro-5-fluorobenzene-1,2-diamine
C6H6ClFN2
MFCD00042485
735773
NC1=CC(F)=C(Cl)C=C1N

Specifications

110°C to 114°C
1 g
MFCD00042485
1,2-diamino-4-chloro-5-fluorobenzene, 4-chloro-5-fluoro-o-phenylenediamine, 4-chloro-1,2-diamino-5-fluorobenzene, 4-chloro-5-fluoro-1,2-phenylenediamine, 1,2-benzenediamine,4-chloro-5-fluoro, 1-chloro-4,5-diamino-2-fluorobenzene, 1,2-benzenediamine, 4-chloro-5-fluoro, 5-chloro-4-fluorobenzene-1,2-diamine, timtec-bb sbb003959, buttpark 44\03-90
NC1=CC(F)=C(Cl)C=C1N
160.58
160.58
4-Chloro-5-fluoro-o-phenylenediamine
139512-70-2
C6H6ClFN2
UN2811
BSMPRJISGCTCDC-UHFFFAOYSA-N
4-chloro-5-fluorobenzene-1,2-diamine
735773
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only