accessibility menu, dialog, popup

UserName

FClNH2NH2

CAS RN 139512-70-2

IUPAC Name: 4-chloro-5-fluorobenzene-1,2-diamine
Synonyms: 4-chloro-5-fluorobenzene-1,2-diamine
Molecular Weight (g/mol): 160.58
Molecular Formula: C6H6ClFN2
InChi Key: BSMPRJISGCTCDC-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(Cl)C=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results