missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5-fluoro-o-phenylenediamine, 98%

Catalog No. AAA1395303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1395303 1 g
AAA1395306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1395303 Supplier Thermo Scientific Chemicals Supplier No. A1395303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 139512-70-2 | C6H6ClFN2 | 160.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 139512-70-2
Molecular Formula C6H6ClFN2
Molecular Weight (g/mol) 160.58
MDL Number MFCD00042485
InChI Key BSMPRJISGCTCDC-UHFFFAOYSA-N
PubChem CID 735773
IUPAC Name 4-chloro-5-fluorobenzene-1,2-diamine
SMILES NC1=CC(F)=C(Cl)C=C1N

Specifications

Melting Point 110°C to 114°C
Quantity 1 g
UN Number UN2811
Synonym 1,2-diamino-4-chloro-5-fluorobenzene, 4-chloro-5-fluoro-o-phenylenediamine, 4-chloro-1,2-diamino-5-fluorobenzene, 4-chloro-5-fluoro-1,2-phenylenediamine, 1,2-benzenediamine,4-chloro-5-fluoro, 1-chloro-4,5-diamino-2-fluorobenzene, 1,2-benzenediamine, 4-chloro-5-fluoro, 5-chloro-4-fluorobenzene-1,2-diamine, timtec-bb sbb003959, buttpark 44\03-90
Formula Weight 160.58
Percent Purity 98%
Chemical Name or Material 4-Chloro-5-fluoro-o-phenylenediamine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only