missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine, 96%

Catalog No. aah3326003
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
5 g
Catalog No. Quantity
AAH3326003 1 g
AAH33260MD 250 mg
AAH3326006 5 g
3 options

Catalog No. AAH3326003

Supplier: Thermo Scientific Chemicals H3326003

Only null left

CAS: 331761-46-7 | C12H6Cl2N2S | 281.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 331761-46-7
Molecular Formula C12H6Cl2N2S
Molecular Weight (g/mol) 281.154
MDL Number MFCD01038375
InChI Key QDMQHIKVIOWCGY-UHFFFAOYSA-N
Synonym 4-chloro-5-4-chlorophenyl thieno 2,3-d pyrimidine, 4-chloro-5-4-chloro-phenyl-thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-4-chlorophenyl, 4-chloro-5-4-chlorophenyl thiopheno 2,3-d pyrimidine, 4-chloranyl-5-4-chlorophenyl thieno 2,3-d pyrimidine
PubChem CID 708755
IUPAC Name 4-chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine
SMILES C1=CC(=CC=C1C2=CSC3=C2C(=NC=N3)Cl)Cl

Specifications

Melting Point 139.5°C to 141°C
Quantity 1 g
Formula Weight 281.16
Percent Purity 96%
Chemical Name or Material 4-Chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only