CAS RN 331761-46-7
IUPAC Name:
4-chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine
Synonyms:
4-chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine
Molecular Weight (g/mol):
281.15
Molecular Formula:
C12H6Cl2N2S
InChi Key:
QDMQHIKVIOWCGY-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1)C1=CSC2=NC=NC(Cl)=C12
1
–
1
of
1
results
1
Promotions Available
4-Chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine, 96%
4-Chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine, 96%
CAS: 331761-46-7 Molecular Formula: C12H6Cl2N2S Molecular Weight (g/mol): 281.154 MDL Number: MFCD01038375 InChI Key: QDMQHIKVIOWCGY-UHFFFAOYSA-N Synonym: 4-chloro-5-4-chlorophenyl thieno 2,3-d pyrimidine,4-chloro-5-4-chloro-phenyl-thieno 2,3-d pyrimidine,thieno 2,3-d pyrimidine,4-chloro-5-4-chlorophenyl,4-chloro-5-4-chlorophenyl thiopheno 2,3-d pyrimidine,4-chloranyl-5-4-chlorophenyl thieno 2,3-d pyrimidine PubChem CID: 708755 IUPAC Name: 4-chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine SMILES: C1=CC(=CC=C1C2=CSC3=C2C(=NC=N3)Cl)Cl