Learn More
4-Chloro-3-(trifluoromethyl)pyridine hydrochloride, 97%
CAS: 732306-24-0 | C6H4Cl2F3N | 218.00 g/mol
Supplier: Thermo Scientific Chemicals L19556MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Chloro-3-(trifluoromethyl)pyridine hydrochloride | |
| ∼135°C (sublimation) | |
| MFCD04972770 | |
| 4-chloro-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethyl pyridinehydrochloride, 4-chloro-3-trifluoromethyl pyridine, chloride, 4-chloro-3-trifluoromethyl pyridine hcl, 4-chloro-3-trifluoromethylpyridine hydrochloride, 3-chloro-4-trifluoromethyl pyridine hydrochlrorid, 4-chloranyl-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethylpyridine hydrochloride salt | |
| Cl.FC(F)(F)C1=C(Cl)C=CN=C1 | |
| 218.00 | |
| 218.01 |
| 732306-24-0 | |
| C6H4Cl2F3N | |
| 250 mg | |
| HCKXJUZITVCDND-UHFFFAOYSA-N | |
| 4-chloro-3-(trifluoromethyl)pyridine hydrochloride | |
| 2782815 | |
| 97% |
Chemical Identifiers
| 732306-24-0 | |
| 218.00 | |
| HCKXJUZITVCDND-UHFFFAOYSA-N | |
| 2782815 | |
| Cl.FC(F)(F)C1=C(Cl)C=CN=C1 |
| C6H4Cl2F3N | |
| MFCD04972770 | |
| 4-chloro-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethyl pyridinehydrochloride, 4-chloro-3-trifluoromethyl pyridine, chloride, 4-chloro-3-trifluoromethyl pyridine hcl, 4-chloro-3-trifluoromethylpyridine hydrochloride, 3-chloro-4-trifluoromethyl pyridine hydrochlrorid, 4-chloranyl-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethylpyridine hydrochloride salt | |
| 4-chloro-3-(trifluoromethyl)pyridine hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only