accessibility menu, dialog, popup

UserName

CAS RN 732306-24-0

IUPAC Name: 4-chloro-3-(trifluoromethyl)pyridine hydrochloride
Synonyms: 4-chloro-3-(trifluoromethyl)pyridine hydrochloride
Molecular Weight (g/mol): 218
Molecular Formula: C6H4Cl2F3N
InChi Key: HCKXJUZITVCDND-UHFFFAOYSA-N
SMILES: Cl.FC(F)(F)C1=C(Cl)C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results